VWR®, Disposable Weighing Dishes
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
25433-014 25433-020
611-1373EA 47.4 GBP
611-1373 611-1374 611-1375 611-1376 611-1377 25433-014 25433-020
VWR®, Disposable Weighing Dishes
Boats Weighing Boats
Aluminium, light gauge.
  • Flanged edge with curled lip for increased stability
  • Contaminant- and oil-free
  • Stackable
Ideal for weighing, sampling, dispensing, storage, food processing, evaporating or general laboratory use.