VWR®, Disposable Weighing Boats
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-0093EA 50.4 GBP
611-0093 611-0094 611-0095 611-9168 611-9169 611-9170 611-9175 611-9176 611-9177 611-9178 611-9179 611-9180 611-9181 611-9182 611-9183 611-3113 611-3114 611-3115
VWR®, Disposable Weighing Boats
Boats Weighing Boats
PS, square.
  • Shallow form with wide, flat bottom avoids tipping
  • Rounded corners simplify transfer
  • Temperature resistant up to 70 °C
These flexible weighing boats, with smooth surfaces, enable accurate pour-out with minimal sample loss. They are biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bleaches. Ideal for use with microwaves, balances or moisture analysers.
Packaging: These weighing boats are produced and packed in plastic bags in a cleanroom.