Hydrochloric acid 36 - 38%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

6081.2500EA 57.9 GBP
6081.2500 6081.1000 6081.9025 6081.2500PE
Hydrochloric acid 36 - 38%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Hydrochloric acid
Formula: HCl
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 37 - 38 %
Ammonium (NH4) max. 3 ppm
Mercury (Hg) max. 5 ppb
Free Chlorine max. 0.5 ppm
Residue after Ignition max. 5 ppm
Sulfate (SO4) max. 1 ppm
Sulfite (SO3) max. 2 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.01
Aluminium (Al) max. 0.05
Arsenic (As) max. 0.05
Barium (Ba) max. 0.02
Beryllium (Be) max. 0.01
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.02
Cobalt (Co) max. 0.01
Chromium (Cr) max. 0.05
Copper (Cu) max. 0.01
Iron (Fe) max. 0.20
Germanium (Ge) max. 0.1
Potassium (K) max. 0.10
Lithium (Li) max. 0.01
Magnesium (Mg) max. 0.05
Manganese (Mn) max. 0.01
Molybdenum (Mo) max. 0.02
Sodium (Na) max. 0.30
Nickel (Ni) max. 0.05
Lead (Pb) max. 0.05
Strontium (Sr) max. 0.01
Titanium (Ti) max. 0.05
Thallium (Tl) max. 0.05
Vanadium (V) max. 0.01
Zinc (Zn) max. 0.5
Zirconium (Zr) max. 0.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR