Silica gel, Pore size: 60 Å 70-230 mesh, high purity for column chromatography, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5


M-Clarity™ quality level = MQ100

60741-1KG 60741-25KG 60741-5KG
60741-1KGEA 126 GBP
60741-1KG 60741-25KG 60741-5KG
Silica gel, Pore size: 60 Å 70-230 mesh, high purity for column chromatography, Supelco®
Silica gel
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=269b27dcefc3b5c7fb165351f35e8d8c78e597a9&vid=b12160b7bd956d4812c0c9d911a65dbdcfeefa50&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR