Potassium hexafluorosilicate, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potassium fluorosilicate

MQ100

60322-1KG
60322-1KGEA 66.2 GBP
60322-1KG
Potassium hexafluorosilicate, Supelco®
Potassium hexafluorosilicate
Formula: F₆K₂Si
MW: 220.27 g/mol
Density: 2.67 g/cm³ (20 °C)
MDL Number: MFCD00040666
CAS Number: 16871-90-2
EINECS: 240-896-2
UN: 2655
ADR: 6.1 III
Merck Index: 12,07803
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d03c363a97c604793b24d9720a51c992c524fd57&vid=c5330082cd3e6f964c9122e98f18213657e04411&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR