Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Potash

60110-1KG 60110-5KG 60110-20KG
60110-1KGEA 80.2 GBP
60110-1KG 60110-5KG HONC60110-20KG
Potassium carbonate, anhydrous ≥99.0% (by titrimetric analysis), Purum p.a.
Potassium carbonate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: K₂CO₃
MW: 138.21 g/mol
Boiling Pt: Dec.
Melting Pt: 891 °C
Density: 2.430 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR