Thiamine hydrochloride, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

5871-100
5871-100EA 71.9 GBP
5871-100
Thiamine hydrochloride, Millipore®
Thiamine hydrochloride
Formula: C₁₂H₁₈Cl₂N₄OS
Melting Pt: 252 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR