Calbiochem® recombinant tetanus toxoid
Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
582231-25 582231-100
582231-25EA 267 GBP
582231-25 582231-100
Calbiochem® recombinant tetanus toxoid
Proteins and Peptides
This tetanus toxoid is prepared by formaldehyde inactivation of tetanus toxin.
It retains its antigenic character, but is essentially non toxic as determined by an LD₅₀ in mice.