Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulpho-SMCC, Sulfo-SMCC

M-Clarity™ quality level = MQ100

573115-50
573115-50EA 190 GBP
573115-50
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) ≥94% (by HPLC), Calbiochem®, Millipore®
Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

SMCC, Water-Soluble, a water-soluble derivative of SMCC.


Ideal for coupling enzymes to antibodies. Contains a cyclohexane bridge, which gives extra stability to the maleimide group. Both enzyme activity and antibody specificity are preserved after coupling. Water-soluble cross-linking reagents help to avoid the use of organic solvents such as dimethylformamide (DMF) and dimethylsulfoxide (DMSO) when preparing bioconjugates.


Ideal for coupling enzymes to antibodies. Contains a cyclohexane bridge, which gives extra stability to the maleimide group. Both enzyme activity and antibody specificity are preserved after coupling.

Formula: C₁₆H₁₈N₂O₉S
Storage Temperature: Freezer
MDL Number: MFCD00054978
CAS Number: 92921-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR