Sodium salicylate, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

Synonyms: Sodium 2-hydroxybenzoate, 2-Hydroxybenzoic acid sodium salt, Salicylic acid sodium salt

567630-5
567630-5EA 47.5 GBP
567630-5
Sodium salicylate, Millipore®
Sodium salicylate
Formula: HOC₆H₄COONa
MW: 160.10 g/mol
Melting Pt: 200
Density: 0.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002440
CAS Number: 54-21-7
EINECS: 200-198-0
Merck Index: 12,08819

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR