Potassium hydroxide 0.1 N ethanol, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: Potash caustic, Caustic potash

M.O.C. Category N

5645.1000EA 74.6 GBP
5645.1000
Potassium hydroxide 0.1 N ethanol, BAKER ANALYZED® Volumetric, J.T.Baker®
Potassium hydroxide

Volumetric Solution, ready for use


Each lot of this product is standardized potentiometrically against an NIST Standard Reference Material, Potassium Hydrogen Phthalate.

Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 II
Merck Index: 12,07806

Order Now

Specification Test Results

Titer (mol/l) 0.0995-0.1005
A light red color can occur in time due to product characteristics Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR