Dodecamolybdophosphoric acid hydrate ACS

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

56166.22 56166.06 56166.14 56166.36
56166.22EA 108 GBP
56166.22 56166.06 56166.14 56166.36
Dodecamolybdophosphoric acid hydrate ACS
Dodecamolybdophosphoric acid hydrate

CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Physical Form: Granular Density: 3.15 Sensitivity: Moisture Sensitive Appearance: Yellow Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913

Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
Melting Pt: 78…90 °C
Density: 3.15
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3085
ADR: 5.1,III
Merck Index: 14,7342

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR