Anti-MAPK9 Rabbit Polyclonal Antibody

Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

559304-200
559304-200EA 492 GBP
559304-200
Anti-MAPK9 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAPK9 (mitogen-activated protein kinase 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR