Consumables for PerkinElmer autosamplers
Supplier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
000475 000478 001953 001954 001955 001957 004670 033750 033765 072624 0726280 092091 092092 092095 092097 092100 092103 092236 092244 0726536 092098 092101 09209225 09209525 0920990 09210001 09210101 09210301 09210025 09209501 09224401 0726522
549-0397EA 123.8 GBP
549-0397 562-1003 562-1018 562-1019 562-1020 562-1021 549-0307 549-0508 549-0509 558-0353 558-0354 558-0291 562-1089 558-0292 558-0463 558-0294 558-0296 558-0297 558-0298 558-0418 558-0293 558-0295 558-0460 558-0498 558-0591 SGEA09210001 SGEA09210101 SGEA09210301 558-0593 SGEA09209501 SGEA09224401 558-0597
Consumables for PerkinElmer autosamplers
Chromatography Supplies Chromatography Spare Parts
Autosampler syringes for PerkinElmer AutoSystem.
* Gas-tight
** Denotes spare needle and plunger kit
Order Now
SPECIFICATIONS
Environmentally Preferable