Rifampicin, Millipore®

Supplier: Merck Millipore Calbiochem
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP

557303-1 557303-5
557303-1EA 80.1 GBP
557303-1 557303-5
Rifampicin, Millipore®
Rifampicin
Formula: C₄₃H₅₈N₄O₁₂
Melting Pt: 183…188 °C
MDL Number: MFCD00151389
CAS Number: 13292-46-1
EINECS: 236-312-0
Merck Index: 12,08382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR