VWR® Hichrom Precolumn Filters
Supplier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
554-7720EA 66.7 GBP
554-7720 554-7719 HI-685 554-7722 HI-565 HI-615 HI-686 HICHHI-602X
VWR® Hichrom Precolumn Filters
Filters Precolumn Filters and Frits
Placed immediately before the column, the precolumn filter traps particles introduced with the sample. For samples, which may irreversibly absorb onto the column, the use of guard cartridges is strongly recommended.
  • Protect columns from particulates, ultra low dead volume
  • Virtually no dead volume with no band broadening or loss of efficiency
  • Compatible with all HPLC columns
Filter HI-704 comprises a stainless steel holder with a 0,020" thru hole and a 2 µm PEEK encapsulated stainless steel frit (supplied).
The fingertight HI-685 precolumn filterMore Product Information