Micro-metering valves, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
P-445 P-446 P-447
UPCHP-445EA 170 GBP
UPCHP-445 554-3114 UPCHP-447
Micro-metering valves, Upchurch Scientific®
Valves Micro Valves
For fine control of fluid flow rates, these valves can reduce outgoing flow. These needle valves are perfect for use with peristaltic pump fluid transfer applications, mass spectrometry and fraction collection. They can also be used to regulate gas flow in helium sparging lines and as a flow-dependent variable back pressure regulator.
  • Flow rates as low as 3,5 µl/min