Rapid replacement Pods™, Rheodyne®
Supplier: IDEX RHEODYNE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PD715-105 PD7900 PD7970 PD9900 PD9960
554-2676EA 2120 GBP
554-2676 554-2677 554-2678 554-2680 554-3297
Rapid replacement Pods™, Rheodyne®
Chromatography Supplies Chromatography Spare Parts
The Rapid Replacement Pod™ (removable liquid-end) design of the TitanHP™ and TitanHT™ platforms make it easy to replace valve's wear components. The entire Pod™ can be purchased separately and replaced as a unit with nearly no instrument downtime for repair. The Pods™ can then be rebuilt at a later time with genuine Rheodyne® RheBuild® kits.
  • Rapid replacement for nearly zero down time
  • Extended valve life