One-piece fingertight fittings
Supplier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
JR-M55020-1 JR-M55021-1 JR-M55022-1 JR-M55023-1 JR-M55024-1 JR-M55025-1 JR-M5502
554-1070EA 92.8 GBP
554-1070 554-1147 554-1148 554-1149 554-1150 554-1151 554-1069
One-piece fingertight fittings
Fittings and Connectors
PEEK one-piece fingertight fittings. Work with all ¹/₁₆'' OD tubings and female 10 to 32 ports. They are rated to 350 bar and can be used with virtually any HPLC fittings, eg. Swagelok, Parker, Rheodyne, Valco.
  • 100% inert and biocompatible
  • Same colour coding as capillary tubing