Twin trough chambers
Supplier: CAMAG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
022.5155 022.5156 022.5253 022.5254 022.5255 022.5256 022.5258
552-0011EA 280 GBP
552-0011 552-0012 552-0018 552-0019 552-0020 552-0021 552-0023
Twin trough chambers
TLC Development Tanks
Twin-trough chambers offer several ways to improve the results of TLC/HPTLC developing techniques.
  • Low solvent consumption
  • Reproducible pre-equilibration of the plate with solvent vapour is possible by placing it in the empty trough opposite that containing the solvent
Development only starts when the solvent is introduced into the trough with the plate.
* ADC: automatic developing chamber.