VWR®, TLC Glass Plates, Silica HP, Unmodified Layers

Supplier: VWR Collection
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

551-0007EA 174 GBP
551-0007 551-0012 551-0015 551-0016 551-0025
VWR®, TLC Glass Plates, Silica HP, Unmodified Layers
TLC Plates
These TLC glass plates have an excellent separation efficiency due to an optimised particle size distribution.

  • Specific dyeability and abrasion resistance based on an optimised binder system
  • Good wettability for exact colorisation results, even with 100% aqueous detection reagents

Specific surface (BET): ~500 m²/g

Specific pore volume: 0,75 ml/g

Particle size: 5 - 17 µm
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR