VWR®, Micro Syringes Ultra-minute Volume
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
549-0198EA 51 GBP
549-0198 549-0197
VWR®, Micro Syringes Ultra-minute Volume
Syringes Syringes for Chromatography
Micro syringe with fixed needle for high sensitivity GC analysis and ultra-minute volumes.
  • Zero dead volume between plunger and needle
  • Fixed needle with bevelled tip for smooth septum penetration
Length in ordering table refers to needle length.