VWR® PFAS Kit
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
548-7011AEA 38.4 GBP
548-7011A 548-7008A
VWR® PFAS Kit
Vials
Designed for for PFAS analysis, these kits contain 0,7 ml polypropylene snap ring vials with silicone white/polypropylene transparent septum, available with or without slit.
  • PFAS are highly persistent and are therefore also known as perpetual chemicals; they are toxic and harmful to health
  • More than 1,000 chemicals belong  to this group of substances
  • Many everyday objects contain these problematic substances, which enter the human body either directly or via the environment
  • The fluorine compounds are hardly degradable and therefore remain in the
More Product Information