XCHANGE® syringes
Supplier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2926210 2926230 2928310 2928330 2928410 2928430 2928510 2928530 2928610 2928630 2928730 2928820 2928830 2928210
548-1805EA 97 GBP
548-1805 548-1806 548-1807 548-1808 548-1809 548-1810 548-1811 548-1812 548-1813 548-1814 548-1816 548-1817 548-1818 SGEA2928210
XCHANGE® syringes
Syringes Syringes for Chromatography
The XCHANGE® coupling system secures and locks the syringe in place. This is achieved by a coupling mechanical piece that captures the syringe and a magnet that automatically aligns – eliminating the need for springs, screws or alignment gauges. Once engaged the syringe is locked into position requiring zero holding force.
  • Injection of small and large volumes in one sample sequence
  • Adding of internal standards without cross-contamination
  • Preventative exchange of syringe when using harmful media
Order Now
SPECIFICATIONS
Environmentally Preferable