VWR®, Septa ND11
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
548-3302AEA 190 GBP
548-3302A 548-0396A 548-0400A 548-0393A 548-0394A 548-0398A 548-0397A 548-0184A 548-0395A 548-0843A 548-1554A 548-2516A
VWR®, Septa ND11
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.