Recombinant protein G, peroxidase conjugated, Calbiochem®
Supplier: Merck Millipore (Oncogene)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
539322-500
539322-500EA 208 GBP
539322-500
Recombinant protein G, peroxidase conjugated, Calbiochem®
Protein Purification Protein Purification Reagents
Purified recombinant Protein G conjugated to peroxidase can be used in direct immunoassay.
  • Used in enzyme-linked immunosorbent assay and immunoblotting (Western blotting)
  • Binds ≥99% to human IgG immobilised on a column