Incinerating dishes, rectagular
Supplier: MORGAN ADVANCED MATERIALS HALD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AL/50X38X36 ALGK50X20X20 ALGK50X25X20 ALGK50X38X36 ALGK75X50X25 ALGK100X32X28 ALGK100X45X19 ALGK105X15X15 ALGK150X65X19 ALGK150X65X35 ALGK160X135X90 ALGK160X80X30 ALGK40X10X8 LGK/AL7/50X25 LGK/AL7/100X45
HALDAL/50X38X36EA 148 GBP
HALDAL/50X38X36 539-1924 HALDALGK50X25X20 HALDALGK50X38X36 HALDALGK75X50X25 HALDALGK100X32X28 HALDALGK100X45X19 HALDALGK105X15X15 HALDALGK150X65X19 HALDALGK150X65X35 HALDALGK160X135X90 HALDALGK160X80X30 HALDALGK40X10X8 HALDLGK/AL7/50X25 HALDLGK/AL7/100X45
Incinerating dishes, rectagular
Boats Combustion Boats
Alsint 99,7.
  • Good thermal shock resistance due to high thermal conductivity
  • Resistant to chemical attack, e.g. hydrogen and other reducing gases