Dishes, crystallising, quartz
Supplier: QUARTZTECHNISCHE WERKSTATTEN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1133 1134 1135 1136 1137 1130 1124
539-1852EA 59.6 GBP
539-1852 539-1854 539-1856 539-1858 539-1860 QUTW1130 QUTW1124
Dishes, crystallising, quartz
Trays/Dishes/Pots Crystallising Dishes
Quartz glass, clear.
  • Extremely low expansion
  • Best possible elastricity and thermal shock resistance
  • High transformation and softening point temperature
  • Low heat conductivity
  • Low electrical conductivity
For a wide range of applications.