2-Butoxyethanol ≥99%

Supplier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Warning

537551-1L
537551-1LEA 64.2 GBP
537551-1L
2-Butoxyethanol ≥99%
2-Butoxyethanol
Formula: CH₃(CH₂)₃OCH₂CH₂OH
Boiling Pt: 168…172 °C (1.013 hPa)
Melting Pt: –74.8 °C
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 10.6 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00002884
CAS Number: 111-76-2
EINECS: 203-905-0
ADR: 6.1,III
Merck Index: 13,01558
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba5a9d2c527d0c167d2248d1936e2d1bd4aeab7a&vid=75260b2608ddc5eb29642178ab193fbcaf54198d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

IR spectrum complying
Assay (GC) Min. 99.0 %
Density (20°C) 0.9000 - 0.9040 g/ml
Water (Karl Fischer) Max. 0.10 %
Free acid (as CH₃COOH) Max. 0.010 %
APHA Max. 10
Vapor Pressure <1 mmHg ( 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR