Freeze dryer, bench top, FreeZone®
Supplier: LABCONCO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
700201050 700202050 700401050 700402050 700801050 700802050 710201050 710202050 710401050 710402050 720401050 720402050 710202015
535-0394EA 0 GBP
535-0394 535-0396 535-0398 535-0400 535-0389 535-0390 535-0402 535-0404 535-0406 535-0408 535-0414 535-0415 LBCP710202015
Freeze dryer, bench top, FreeZone®
Freeze Dryers
FreeZone® bench top freeze dryers are designed for lyophilising light to moderate sample loads. The compact design is ideal for laboratories with limited floor space.
  • Consistent, high quality results with flexible programming, and on board data collection
  • Challenging samples are easy with accessories designed specifically for laboratory samples
  • Brushed stainless steel and glacier white, powder-coated steel exterior with blue accents
  • End-Zone™ end point detection option takes the guess work out of sample dryness
  • PTFE-coated collector coil and chamber for
More Product Information