VWR® Separating Funnels, Squibb
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
532-0018EA 64.6 GBP
532-0018 532-0029 532-0019 532-0020 532-0021 532-0022 532-0023 532-0024 532-0025 532-0026 532-0027 532-0028
VWR® Separating Funnels, Squibb
Funnels Separating Funnels
Borosilicate 3.3 glass with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance