VWR®, Separating Funnels, Conical
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
532-0007EA 65.9 GBP
532-0007 532-0008 532-0009 532-0010 532-0011 532-0012 532-0013 532-0014 532-0015 532-0006 532-0017 532-0016
VWR®, Separating Funnels, Conical
Funnels Separating Funnels
Borosilicate 3.3 glass, with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance