Rotary evaporator receiving flasks
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
03063558 03063570 03063573 03063537 03063549 03064037 03064049 03064058 03064070 03064073 03064076 03064079
531-9144EA 119 GBP
531-9144 531-9145 531-9146 531-9142 531-9143 531-0825 531-0826 531-0827 531-0821 531-0822 531-0823 531-0824
Rotary evaporator receiving flasks
Flasks Rotary Evaporator Flasks
DURAN®, borosilicate glass, clear or plastic-coated.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Spherical joint KS 35/20, centred, suitable for all rotary evaporators.