VWR® Glassware and Accessories for Rotary Evaporators, RV 10/RV 8
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3655300 3755300 3755400 3755500 3741000 3741100 3742000 3742100 3743000 3743100 3744000 3744100 3812200 3739200 3739400 3739500 3739600 3740100 3740200 3740300 3740400 3740500 3740600 3740700 3740800 3740900 3742200 3742300 3742400 3742500 3742600 3742700 3743200 3738200 3738300 3738400 3738800 3738900 3743300 3743400 3743500 3743600 3743700 3903800 3859000 3907100 3741200 3741300 3919400 3971700 3907000 4221400 20005330 20005331 20005332 20005333 20005334 20005335 3638900 20019654
531-3476EA 1290 GBP
531-3476 531-3477 531-3478 531-3479 531-0460 531-0461 531-0462 531-0463 531-0464 531-0465 531-0466 531-0467 531-0468 531-3000 531-3001 531-3002 531-3003 531-3004 531-3005 531-3006 531-3007 531-3008 531-3009 531-3010 531-3011 531-3012 531-3013 531-3014 531-3015 531-3016 531-3017 531-3018 531-3019 531-3025 531-3026 531-3027 531-3028 531-3029 531-3020 531-3021 531-3022 531-3023 531-3024 531-3489 531-0672 531-0673 531-0674 531-0675 531-0676 531-1033 531-0842 531-1032 IKAA20005330 IKAA20005331 IKAA20005332 IKAA20005333 IKAA20005334 IKAA20005335 IKAA3638900 IKAA20019654
VWR® Glassware and Accessories for Rotary Evaporators, RV 10/RV 8
Evaporators Rotary Evaporators
Glassware sets include 1000 ml evaporation and receiving flasks. The solvent being distilled can be continuously fed in through the PTFE inlet tube. Plastic coated versions are available and offer additional user protection.
Other options include a heating bath shield for optimal protection against splashes of hot liquid. A protective cover which is essential when it is not possible to work in an extractor hood (protects the user against splashes of hot liquid and in the event of the evaporator flask breaking - coated glassware should also be used).
* With transparent plastic coating.