Immersion thermostats, IC series
Supplier: IKA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3861000 3863000
531-1061EA 3960 GBP
531-1061 531-1062
Immersion thermostats, IC series
Circulating Thermostats Open Circulator Systems Thermostat Controllers
The IC immersion circulators have an extended temperature range for tempering fluids in open and covered water baths up to 200/250 °C. The LED/ graphic TFT Display illustrates all relevant process parameters. Units feature push and turn buttons for intuitive menu navigation. IC immersion circulators ensure short heating up times and have a temperature stability of ±0,01/0,02 K (basic/control model). Both models feature optical and acoustic safety warnings, an adjustable safety circuit, a powerful pressure/suction pump and can be used for internal and external applications.