Separating funnels, Squibb, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4300-0125 4300-0250 4300-0500 4300-1000
527-0005EA 162 GBP
527-0005 527-0006 527-0007 527-0008
Separating funnels, Squibb, Nalgene®
Funnels Separating Funnels
PPCO translucent, TFE stopcock, PP screw closure.
  • Leakproof closure and stopcock
  • Very good chemical resistance
  • Autoclavable except for stopcock assembly
  • TFE stopcock requires no lubrication