VWR®, Ice-Free Cooling System for PCR Preparation and Storage
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
525-1489EA 137 GBP
525-1489 525-1491 525-1490
VWR®, Ice-Free Cooling System for PCR Preparation and Storage
Boxes Cooling/Freezing Boxes and Bags
This robust cooler is consistent and reliable. It is made from engineered PP with a PC lid. The modern design cools faster in freezers and stays cold longer on the bench top.
  • Better alternative for ice baths as dry incubation reduces the risk of contamination
  • Freeze cooler in an upright position enables cold storage of tubes
  • Well design prohibits the freezing of samples
  • Fully sealed and sonic welded cooler keeps coolant contained offering worry-free usage
  • Saves space with compact footprint and stacking ability
  • The lid is ambidextrous; it can be switched to
More Product Information