n-Hexacosane, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synonyms: N-hexacosane

MQ100

52183-1G 52183-250MG
52183-1GEA 207 GBP
52183-1G 52183-250MG
n-Hexacosane, Supelco®
n-Hexacosane
Formula: H₃C(CH₂)₂₄CH₃
MW: 366.71 g/mol
Boiling Pt: 412.2
Melting Pt: 56…59 °C
Density: 0.804 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009354
CAS Number: 630-01-3
EINECS: 211-124-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec0fc9961cce69c08f514a2cad4ea2dc785dc967&vid=d098f96663d77496576f54cc2e20bf863284bdcf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR