Fixed-angle titanium rotor package/assembly, TLA-120.2
Supplier: Beckman Coulter
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
357656 362046
521-3914EA 0 GBP
521-3914 521-3969
Fixed-angle titanium rotor package/assembly, TLA-120.2
Centrifuges Rotors
Major applications: Pelleting of subcellular fractions in 5 to 30 minutes, plasmid DNA separations in 3 hours.
Note: Non-precipitating solutions up to 1,7 g/ml in density can be run in this rotor without a reduction in rotor speed.