Bench top centrifuges and packages, Megafuge ST1 and ST1R Plus
Supplier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
75009230 75009270 75009730 75009770 75016026 75016019 75017019 75016025 75016003 75017003
521-3433EA 4690 GBP
521-3433 521-3435 521-3437 521-3439 521-3444 521-3446 521-3447 521-3449 521-3457 521-3458
Bench top centrifuges and packages, Megafuge ST1 and ST1R Plus
Centrifuges Benchtop Centrifuges
Thermo Scientific™ Megafuge ST1/ST1R Plus bench top centrifuges offer intuitive controls and versatility for power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
Biocontainment solution seals with a snap; eliminates screw capsMore Product Information