Bench top centrifuges and packages, Multifuge X1 and X1R Pro
Supplier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
75009210 75009250 75009710 75009750 75016024 75017024 75016023 75017023 75016095 75016097
521-3432EA 5940 GBP
521-3432 521-3434 521-3436 521-3438 521-3443 521-3445 521-3448 521-3450 521-4281 521-4282
Bench top centrifuges and packages, Multifuge X1 and X1R Pro
Centrifuges Benchtop Centrifuges
Ergonomically designed, the Thermo Scientific™ Multifuge X1/X1R Pro series of bench top centrifuges have a touch screen interface that enables easier programming for quicker results, and the ability to access and track all runs, rotor life and centrifuge health information for maximum versatility and power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switchingMore Product Information