VWR®, Qualitative Filter Papers
Supplier: VWR Collection
Ratings:
Avg. Ratings: 3.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
516-0800EA 12.7 GBP
516-0800 516-0801 516-0802 516-0803 516-0804 516-0805 516-0820 516-0821 516-0822 516-0823 516-0824 516-0825 516-0826 516-0827 516-0828 516-0829 516-0830 516-0831 516-0832 516-0833 516-0834 516-0835 516-0836 516-0837 516-0806 516-0807 516-0808 516-0809 516-0810 516-0811 516-0812 516-0813 516-0814 516-0815 516-0816 516-0817 516-0818 516-0819 516-0838 516-0839 516-0840 516-0841 516-0842 516-0843 516-0844 516-0845 516-0880 516-0307 516-0276 516-0277 516-0278 516-0279 516-0280 516-0281 516-0282 516-0284 516-0285 516-0286 516-0287 516-0288 516-0290 516-0291 516-0292 516-0293 516-0297 516-0298 516-0300 516-0301 516-0309 516-0283 516-0289 516-0294 516-0295 516-0296 516-0299 516-0302 516-0303 516-0304 516-0305 516-0306 516-0308 516-0310 516-0349 516-0351 516-0350
VWR®, Qualitative Filter Papers
Filters Filter Papers Qualitative Filter Paper
Grade 402 and 302: General grade for qualitative tasks. Suitable for most sulphides. Determination of total fat content in dairy products.
Grade 403 and 313: Drugs, pharmaceuticals, liquors, tinctures, chromatography and protein-free filtrations in the clinical laboratory.
Grade 410 and 305: Filtration of copper(I) oxide, lead sulphate, barium sulphate, manganese carbonate and more for analytical work. Suitable for extremely difficult filtration and for clarification of wine.
Grade 413 and 303: For routine laboratory work, suitable for student requirements in general chemistry, also forMore Product Information