n-Heptadecane, Supelco®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Danger

Synonyms: N-heptadecane

MQ100

51578-5ML
51578-5MLEA 0 GBP
51578-5ML
n-Heptadecane, Supelco®
n-Heptadecane
Formula: H₃C(CH₂)₁₅CH₃
MW: 240.47 g/mol
Boiling Pt: 302.1
Melting Pt: 21.2
Density: 0.7783 g/cm³ (20 °C)
Flash Pt: 148
Storage Temperature: Ambient
MDL Number: MFCD00009002
CAS Number: 629-78-7
EINECS: 211-108-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46f19004bc6f19fe92d031d2ab8a5d29634d4ea8&vid=0fda84699d7859b5f1b4834c4d93f1cff7948a7b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR