Vacuum filtration manifold, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DS0345-0001
513-3451EA 1790 GBP
513-3451
Vacuum filtration manifold, Nalgene®
Manifolds
Vacuum filter funnel manifold for use with Nalgene analytical filter funnels and test filter funnels.
  • Made from stainless steel
  • Two-way valve with vent port