Filtration crucibles, VitraPOR®
Supplier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
20082 20083 20084 20151 20152 20153 20154 20301 20302 20304 20305 20501 20502 20503 20504 20505 20155 20080 20081 20150 20300 20500 20085 2050C 2030C 2030F 20303 2050F
511-2102EA 21.9 GBP
511-2102 511-2103 511-2104 511-2201 511-2202 511-2203 511-2204 511-2301 511-2302 511-2304 511-2305 511-2401 511-2402 511-2403 511-2404 511-2405 511-0360 511-0358 511-2101 511-0359 511-0249 511-0361 511-0248 ROBU2050C ROBU2030C ROBU2030F 511-0273 ROBU2050F
Filtration crucibles, VitraPOR®
Crucibles
Filtration crucibles made from borosilicate glass 3.3 with integrated sintered glass filter plate.
Certifications: Porosity 0 to 5 comply with ISO 4793-80. Porosity C (coarse) and F (fine) comply with ASTM E128-94.