Büchner funnels, Hartley pattern
Supplier: DIXON SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
237091001 237091002 237091003 237091004 237091201 237091202 237091203 237091204 237091601 237091602 237091603 237091801 237091802 237091803 237091804
511-0827EA 449 GBP
511-0827 511-0828 511-0829 511-0830 511-0831 511-0832 511-0833 511-0834 511-0837 511-0838 511-0839 511-0840 511-0841 511-0842 511-0843
Büchner funnels, Hartley pattern
Funnels Filtration Funnels Büchner Funnels
Top and base: Borosilicate glass with support plate.
  • Available with choice of perforated disc material
  • Complete with three PE coated clips
  • Available for 4 filter sizes
Three piece construction.