Buchner funnels
Supplier: DIXON SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
232040111 232040112 232040113 232040114 232040121 232040122 232040123 232040124 232040131 232040132 232040133 232040134 232040141 232040142 232040143 232040144
511-0800EA 135 GBP
511-0800 511-0801 511-0802 511-0803 511-0804 511-0805 511-0806 511-0807 511-0808 511-0809 511-0810 511-0811 511-0812 511-0813 511-0814 511-0815
Buchner funnels
Funnels Filtration Funnels Büchner Funnels
Borosilicate glass
  • With sintered glass disc
  • Very good chemical resistance
  • High temperature resistance
For paper or membrane filtration disc.