Filtration funnels, VitraPOR®
Supplier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
22251 22252 22253 22254 22255 22261 22262 22263 22265 22264
511-3301EA 38.1 GBP
511-3301 511-3302 511-3303 511-3304 511-0378 511-0379 511-0380 511-0381 511-0383 511-0382
Filtration funnels, VitraPOR®
Funnels Filtration Funnels Filter Funnels
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.