Micro filter funnels, VitraPOR®
Supplier: ROBU GLASFILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
27200 27080 27084 27201 27202 27203 27204 27081 27082 27083
511-0385EA 183 GBP
511-0385 511-0384 511-0257 ROBU27201 511-0290 511-0291 511-0258 ROBU27081 511-0274 511-0256
Micro filter funnels, VitraPOR®
Funnels Filtration Funnels Micro Filter Funnels
Borosilicate glass 3.3.
  • For filtration of very small volumes
  • Excellent temperature resistance
With integrated sintered glass filter plate.
Order Now
SPECIFICATIONS
Environmentally Preferable