Nylon woven filters, CellMicroSieves™
Supplier: BIODESIGN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N200S N100S N70S N50S N35S N25S N20S N15S N10S N8S NSC N200R N100R N70R N50R N35R N25R N20R N15R N10R N8R N5R N200C N100C N70C N50C N35C N25C N20C N15C N10C N8C N5C
510-0011EA 161 GBP
510-0011 510-0002 510-0033 510-0027 510-0024 510-0017 510-0014 510-0008 510-0005 510-0036 510-0039 510-0010 510-0001 510-0032 510-0026 510-0023 510-0016 510-0013 510-0007 510-0004 510-0035 510-0029 510-0009 510-0000 510-0031 510-0025 510-0022 510-0015 510-0012 510-0006 510-0003 510-0034 510-0028
Nylon woven filters, CellMicroSieves™
Cell Strainers
CellMicroSieves™ allow the rapid, yet gentle, isolation of cells, organelles and cellular debris. Unlike traditional filters, these are a precision woven matrix of pure nylon filaments that are available with pores ranging from 5 to 200 µm. CellMicroSieves™ do not wet and contains no dyes, detergents or extractables. CellMicroSieves™ are biologically inert, autoclavable and can be repeatedly washed and reused. They are resistant to most acidic or basic conditions and many solvents. Extremely low protein affinity means that cells and proteins will not bind to mesh.
  • Rapidly collects
More Product Information